Learn More
Description
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- Hig
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Cross Reactivity | Mouse |
| Includes | Six 96-well PVDF-backed plates coated w/mAb for mouse IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
| Quantity | 6 x 96-well Plates |
| Sample Type | Whole Cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| For Use With (Application) | Diagnostics |
| Product Type | Mouse IFN-gamma ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

