Learn More
R&D Systems™ Mouse IFN-gamma/IL-4 Dual-Color ELISpot Kit
Description
Includes
- Microplate
- Biotin-conjugated Detection Antibody
- HRP-conjugated Detection Antibody
- Streptavidin-conjugated to Alkaline Phosphatase
- Dilution Buffers
- Wash Buffer Concentrate
- BCIP/NBT Chromogen
- AEC
- Detect and quantitate individual cells secreting IFN-γ/IL-4
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
| Includes | One 96-well plate coated w/mAb for mouse IFNG and pAb for mouse IL-4, Biotin- and HRP-conjugated detection Abs, Streptavidin-AP, Buffers, BCIP/NBT, AEC, Positive controls |
| Quantity | 1 x 96-well Plate |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Diagnostics |
| Product Type | Mouse IFNG/IL-4 Dual-Color ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.