Learn More
Specifications
Specifications
| Antigen | IL-2 |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-2 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-2 Positive Control |
| Quantity | 1 x 96-well Plate |
| Sample Type | Whole cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Quantitative determination of the frequency of cells releasing mouse IL-2 |
| Product Type | Mouse IL-2 ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
