Learn More
Description
Includes
- Microplate
- Biotinylated Detection Antibody
- Streptavidin conjugated to Alkaline Phosphatase
- Dilution Buffers
- Wash Buffer Concentrate
- BCIP/NBT Chromogen
- Positive Control
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
| Antigen | IL-4 |
| Conjugate | AP |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-4 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-4 Positive Control |
| Quantity | 6 x 96-well Plates |
| Sample Type | Whole cells |
| Synonym | B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| For Use With (Application) | Detection of cytokine secreting cells, quantitative sandwich ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
