Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse LAG-3 Fc Chimera Protein, CF

Catalog No. 3328L3050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3328L3050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3328L3050 Supplier R&D Systems™ Supplier No. 3328L3050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce TNF-alpha secretion by JAWSII mouse immature dendritic cells. The ED50 for this effect is 0.4-2.4 μg/mL.

Specifications

Accession Number Q61790
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3902
Molecular Weight (g/mol) M.W. (Observed): 85-100 kDa, reducing conditions; M.W. (theoretical): 72.4 kDa (monomer)
Name LAG-3 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse LAG-3 protein Mouse LAG-3 (Gly24-Leu442) (N-terminus) Accession # Q61790 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Lag3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS. Reconstitute 30 minutes prior to use with minimal agitation.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LAG-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.