Description
Includes
Adipogenic Supplement, Mouse/Rat Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP4, Sheep Anti-mouse Collagen II, Goat Anti-mouse Osteopontin
Reduces experimental variation
Reliably induces MSC trilineage differentiation using defined supplements
A panel of antibodies, consisting of Goat Anti-mouse FABP4, Sheep Anti-mouse Collagen II, and Goat Anti-mouse Osteopontin, are included to define th
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |