Description
Includes
25μg each of antibodies to Sca-1, CD106, CD105, CD73, CD29, CD44, CD11b, and CD45
Researchers use different techniques to isolate, culture, and differentiate human mesenchymal stem/stromal cells (MSCs). Variations in experimental approaches as well as differences in the MSC starting population may account for experimental variability a
Cost-effective panel of premium quality antibodies for the positive and negative identification of MSCs
Uses 9 markers to increase confidence in MSC identification
Enables verification of MSC identity in less than 2 hours with use of fluorochrome-conjug
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |