Description
Includes
ITS Supplement, N-2 MAX Supplement, Bovine Fibronectin Stock, Human FGF basic, Human EGF, Human PDGF-AA
Oligodendrocytes are myelinating cells in the central nervous system (CNS) that form the myelin sheath of axons to support rapid nerve conduction. In CNS disorders, such as stroke, multiple sclerosis and spinal cord injury, demyelination of axons contribu
Pre-optimized to reduce experimental variation
Defined conditions decrease variation
High lot-to-lot consistency decreases variation
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |