Learn More
R&D Systems™ Mouse/Rat Eph Sampler Pack 2 Protein Sampler Pack
Description
Regulatory Status: RUO
Specifications
Specifications
| Name | Eph Sampler Pack 2 |
| Quantity | 1 Pack |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.