Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat IL-6 ELISpot Kit
SDP

Catalog No. XEL406 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
XEL406 6 x 96-well Plates
EL406 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. XEL406 Supplier R&D Systems™ Supplier No. XEL406

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

  • The kit is designed for the detection and enumeration of mouse and rat IL-6 secreting cells using an ELISpot assay.
  • Assay Type: Quantitative sandwich ELISA
  • Format: 96-well PVDF-backed microplate
  • Sample Type: Whole cells
  • Sensitivity: High sensitivity; capable of measuring responses with frequencies well below 1 in 100,000 cells
  • Features: Ready-to-run format requiring no assay development
  • Does not require in vitro expansion of cells
  • High-throughput capability using small numbers of primary cells

Specifications

Antigen IL-6
Conjugate AP
Cross Reactivity Mouse
Includes Six 96-well PVDF-backed plates coated w/pAb for mouse/rat IL-6, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Quantity 6 x 96-well Plates
Sample Type Whole Cells
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Gene ID (Entrez) 3569
Gene Symbol Il6
Product Line R&D Systems
For Use With (Application) Diagnostics
Host Species Mouse, Rat
Product Type Mouse/Rat IL-6 ELISpot Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.