Learn More
R&D Systems™ Mouse/Rat Pluripotent Stem Cell Assessment Primer Pair Panel
Description
Includes
mouse/rat ESG1/DPPA5, Nanog, Oct-3/4, SOX2, AFP, GATA-4, PDX-1, SOX17, HNF-3 beta, TP63/TP73L, Otx2, Brachyury, Stella, Nestin, and GAPDH
Stem cells are often defined by the expression of specific marker proteins that differ depending on cell potency and lineage. While no single marker can define a cell population, multiple markers can be used to increase the confidence and accuracy of stem
Verifies differentiation state using PCR
Uses multiple pluripotent and lineage-specific markers: includes 14 primers to amplify stem cell markers, 1 primer to control for cDNA synthesis, and dsDNA to serve as a positive control
Efficient and cost-effect
Specifications
Specifications
| Content And Storage | Store the unopened product at -20° to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 Kit |