Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Serpin F2/alpha 2-Antiplasmin Protein

Catalog No. 1239PI010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

The Recombinant Mouse Serpin F2/alpha 2-Antiplasmin Protein is derived from NS0. The Recombinant Mouse Serpin F2/alpha 2-Antiplasmin Protein has been validated for the following applications: Inhibition Activity.

Specifications

For Use With (Application) Inhibition Assay
Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 18816
Molecular Weight (g/mol) 54 kDa
Name Serpin F2/alpha 2-Antiplasmin
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse Serpin F2/alpha 2-Antiplasmin protein Val28-Lys491,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.