Learn More
R&D Systems™ Mouse Th2 Cell Multi-Color Flow Cytometry Kit
SDP
Catalog No. FMC012
Change view
Click to view available options
25 Tests
Catalog No.
FMC012 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. FMC012 Supplier R&D Systems™ Supplier No. FMC012
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed for the flow cytometric analysis of mouse Th2 cells using four fluorochrome-conjugated antibodies
Single-step stainingIncludes conjugated antibodies and corresponding isotype controls: IL-4 R-CFS (Affinity-purified polyclonal antibody; goat IgG), STAT6-APC (Clone 253906; mouse IgG2A), IL-5-PE (Clone TRFK5; rat IgG1), CD4-PerCP (Clone GK1.5; rat IgG2B)
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.