Learn More
Description
Includes
Mouse Thrombodmodulin Microplate; Mouse Thrombodmodulin Standard; Mouse Thrombodmodulin Control; Mouse Thrombodmodulin Conjugate; Assay Diluent RD1-38; Calibrator Diluent RD5T Concentrate; Wash Buffer Concentrate; Color Reagent A; Color Reagent B; Stop Solution; Plate Sealers
Specifications
Specifications
| Assay Range | 46.90 - 3000pg/mL |
| Assay Sensitivity | 19.1pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Mouse Thrombomodulin |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
