Learn More
R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein. |
| Gene ID (Entrez) | 21943 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | TRANCE/TNFSF11/RANK L |
| Quantity | 10 μg |
| Source | E. coli-derived mouse TRANCE/TNFSF11/RANK L protein Lys158-Asp316,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.