Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Catalog No. p-4830531
Change view
Click to view available options
Quantity:
5000 Tests
Catalog No. Quantity
4890050K 5000 Tests
1 options
This item is not returnable. View return policy
Catalog No. 4890050K Supplier R&D Systems™ Supplier No. 4890050K
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
MTT reagent; Detergent Reagent
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
Quantity 5000 Tests
Product Type MTT Cell Proliferation/Viability Assay
Content And Storage The product requires storage at 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
For Use With (Application) Minimizes steps to assay for proliferation rate in cultured cells