Learn More
R&D Systems™ Multi-species Protein Fraction
Description
Source: Rabbit erythrocytes,
Formulation: Formulation cannot be determined without the BulkLotPrefix
Regulatory Status: RUO
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Quantity | 400 μg |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.