Learn More
R&D Systems™ MycoProbe Mycoplasma Detection Kit
GSA_VASDP
Catalog No. CUL001B Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
CUL001B 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. CUL001B Supplier R&D Systems™ Supplier No. CUL001B
Add to Cart
Add to Cart
This item is not returnable. View return policy
A complete kit to detect common antibiotic-resistant cell culture contaminants
Mycoplasma contamination affects up to 80% of continuous cell cultures. If undetected, mycoplasma contamination can have significant effects on the quality and reliability of cell culture preparations.
Reduces experimental variation
Accurate and highly sensitive
Simple multiwell-based colorimetric assay
Only takes 4.5 hours
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit