Learn More
R&D Systems™ N-2 Plus Media Supplement
SDP
Catalog No. AR003 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
AR003 5 mL
1 options
This item is not returnable. View return policy
Catalog No. AR003 Supplier R&D Systems™ Supplier No. AR003
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
To culture neural progenitor cells and their differentiated derivatives under defined, serum-free conditions
Serum-free, defined media are routinely used as an alternative to standard serum-containing media in order to reduce unwanted experimental variability.
  • Chemically defined and serum-free
  • Consistent composition reduces experimental variation
  • Optimized for neural cell cultures
  • Contains high quality factors to support reproducible and efficient NPC expansion when combined with appropriate growth factors
  • N-2 Plus Media Supplement is a modification of Bottenstein's formulation
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type Media Supplement
Quantity 5 mL