Learn More
R&D Systems™ N21-MAX Insulin Free Media Supplement (50X)
SDP
Catalog No. AR010 Shop All Novus Biologicals Products
Change view
Click to view available options
10mL
Catalog No.
AR010 10mL
1 options
This item is not returnable. View return policy
Catalog No. AR010 Supplier R&D Systems™ Supplier No. AR010
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
The following 20 components for long-term neuronal cell culture: Albumin (bovine); L-Carnitine; Catalase; Corticosterone; Ethanolamine; Glutathione; Galactose; Holo-Transferrin; Linoleic Acid; Linolenic Acid; Lipoic Acid; Progesterone; Putrescine; Retinyl acetate; Retinol; Selenite; Superoxide dismutase; Triiodo-L-thyronine; D,L-alpha-Tocopherol; D,L-alpha-Tocopherol acetate;
Tested in the viability and growth of E18 rat hippocampal neurons
N21-MAX Insulin Free Media Supplement is supplied as a 50X concentrated solution. A serum-free media supplement specially optimized for the long-term culture of neurons.
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 mL