Learn More
Description
- Compatible with cryostat and paraffin-embedded tissue sections, as well as with cultured neuronal and non-neuronal cells
- Protects cells and tissues from shrinking, thus maintaining their native cytological and histological architecture
- Ready-to-use
Applications:
Fluorescence imunohistochemistry and Immunocytochemistry, Fluorescence neuronal tracing, and Fluorescence in situ hybridization
Specifications
Specifications
| Applications | Fluorescence imunohistochemistry and Immunocytochemistry, Fluorescence neuronal tracing, and Fluorescence in situ hybridization |
| Product Type | Guard Mounting Media |
| Content And Storage | Store unopened product at -20°C to -70°C. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
