Learn More
R&D Systems™ ProDots Recombinant Human IL-4
Description
Specifications
Specifications
| Accession Number | P05112 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | Predicted MW: 15kDa Observed MW: 14kDa, reducing conditions |
| Name | ProDots™ Recombinant Human IL-4 |
| Quantity | 50 μg |
| Source | E. coli-derived human IL-4 protein His25-Ser153,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -70° C under sterile conditions after reconstitution |
| Gene Alias | B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.