Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human IL-4

Catalog No. PRD20450
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD20450 250 μg
PRD20410 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD20450 Supplier R&D Systems™ Supplier No. PRD20450
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human IL-4 Protein is derived from E. coli. The ProDots Recombinant Human IL-4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P05112
For Use With (Application) Bioactivity
Molecular Weight (g/mol) Predicted MW: 15kDa Observed MW: 14kDa, reducing conditions
Name ProDots™ Recombinant Human IL-4
Quantity 250 μg
Source E. coli-derived human IL-4 protein His25-Ser153,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -70° C under sterile conditions after reconstitution
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100-200 μg/mL in PBS containing = 0.1% human or bovine serum albumin, or simply roll ProDot™ directly into cell culture medium for immediate use.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.