Learn More
R&D Systems™ Proteome Profiler Mouse Angiogenesis Array Kit
GREENER_CHOICESDP
Catalog No. ARY015 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY015 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY015 Supplier R&D Systems™ Supplier No. ARY015
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Mouse Angiogenesis Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Mouse Angiogenesis Array Kit contains 4 membranes - each spotted in duplicate with 53 different angiogenesis antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit