Learn More
R&D Systems™ Proteome Profiler Rat Adipokine Array Kit
GREENER_CHOICESDP
Catalog No. ARY016 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY016 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY016 Supplier R&D Systems™ Supplier No. ARY016
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Rat Adipokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Rat Adipokine Array Kit contains 4 membranes - each spotted in duplicate with 30 different obesity-related antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit