Learn More
R&D Systems™ Proteome Purify 2 Mouse Serum Protein Immunodepletion Resin
Shop All Novus Biologicals Products
Description
Analysis of low abundance proteins by even the most sensitive of methods for proteomics greatly benefits from the removal of high abundance proteins that may interfere with their detection.
- Improves visualization of target proteins
- Allows user to increase protein load or concentration
Specifications
Specifications
| Quantity | 40 Tests |
| Product Type | Immunodepletion Resin |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Product Line | R&D Systems |