Learn More
Tocris Bioscience™ QAQ Dichloride
Description
Specifications
Specifications
| Chemical Name or Material | QAQ dichloride |
| Synonym | 2,2'-[1,2-Diazenediylbis(4,1-phenyleneimino)bis[N,N,N-triethyl-2-oxo-ethanaminium] dichloride |
| Quantity | 50 mg |
| Solubility | Soluble to 50mM in water |
| Target | Non-selective Cav Blockers |
| Molecular Formula | C28H44Cl2N6O2 |
| Formula Weight | 567.59 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.