Learn More
R&D Systems™ Rat 20S Immunoproteasome Protein
Description
Source: Rat spleen,
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl and DTT.
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Supplied as a solution in HEPES, NaCl and DTT. |
| Quantity | 50 μg |
| Product Type | 20S Immunoproteasome |
| Species | Rat |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.