Learn More
R&D Systems™ Rat CD3+ T Cell Enrichment Columns
Description
Includes
Cell Selection Columns, 10X Column Buffer, Antibody Cocktail Vials
Purifies CD3+ T cells to 85 to 95%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Total CD3+ cell recovery ranges between 30% and 50% of initial CD3+ cells loaded;
Specifications
Specifications
| Quantity | 25/Pk. |
| Product Type | Cell Enrichment Column |