Learn More
Description
Includes
Cell Selection Columns, 10X Column Buffer, Antibody Cocktail Vials
Purifies CD3+ T cells to 85 to 95%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Total CD3+ cell recovery ranges between 30% and 50% of initial CD3+ cells loaded;
Specifications
Specifications
| Quantity | 5/Pk. |
| Product Type | Cell Enrichment Column |
| Product Line | R&D Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
