Learn More
R&D Systems™ Rat CD4+ T Cell Enrichment Columns
SDP
Catalog No. RCD4C1000 Shop All Novus Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
RCD4C1000 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. RCD4C1000 Supplier R&D Systems™ Supplier No. RCD4C1000
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
CD4 Subset Columns, 4; Monoclonal Antibody Cocktail, 4 x 1mL; 10X Column Buffer Concentrate, 30mL
Designed to prepare sub-populations of CD4+ rat T cells via high affinity negative selection
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
Recovery of rat CD3+/CD4+ cells from all available CD3+/CD4+ cells loaded ranged between 40% and 50%
The purity of recovered cells ranged between 89% and 95%, with less than 2% detec
Specifications
Quantity 4/Pk.