Description
Includes
CD8 Subset Columns, 4; Monoclonal Antibody Cocktail, 4 x 1mL; 10X Column Buffer Concentrate, 30mL
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
Recovery of rat CD3+/CD8+ cells from all available CD3+/CD8+ cells loaded ranged between 25% and 35%
The purity of recovered cells ranged between 89% and 95%, with less than 1% detec
Specifications
Specifications
| Quantity | 4/Pk. |
| Product Line | R&D Systems |