Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat EGF Protein

Catalog No. 3214EG100
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
3214EG100 Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
3214EG100CF Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3214EG100 Supplier R&D Systems™ Supplier No. 3214EG100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED50 for this effect is 10-60 pg/mL.

Specifications

Accession Number P07522
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 1950
Molecular Weight (g/mol) M.W. (theoretical): 6.3 kDa
Name EGF
Quantity 100 μg
Source E. coli-derived rat EGF protein Asn974-Arg1026, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Egf
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 10 mM Acetic Acid containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein EGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.