Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat IFN-gamma ELISpot Kit

Catalog No. EL585
Change view
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EL585 1 x 96-well Plate
XEL585 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. EL585 Supplier R&D Systems™ Supplier No. EL585
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells

Specifications

Includes One 96-well PVDF-backed plate coated w/pAb for rat IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT; Positive control
Quantity 1 x 96-well Plate
Sample Type Whole Cells
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Diagnostics
Product Type Rat IFN-gamma ELISpot Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.