Learn More
R&D Systems™ Recombinant Rat IL-6 Protein
Description
Specifications
Specifications
| Accession Number | P20607.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate with BSA as a carrier protein. |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | M.W. (theoretical): 22 kDa |
| Name | IL-6 |
| Quantity | 50 μg |
| Source | E. coli-derived rat IL-6 protein Phe25-Thr211, with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.