Description
Includes
Adipogenic Supplement, Chondrogenic Supplement, Osteogenic Supplement, ITS Supplement; Adipocyte marker: Goat Anti-Mouse FABP4 Ag-affinity Purified Polyclonal Ab; Chondrocyte marker: Goat Anti-Human Aggrecan Ag-affinity Purified Polyclonal Ab; Osteocyte m
Confirms that the starting MSC population is multipotent
Reduces experimental variation
Reliably induces MSC trilineage differentiation using defined supplements
This kit requires media not included, such as Human/Mouse/Rat StemXVivo Osteogenic/Adipoge
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 Kit |