Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat MMP-8 Protein, CF

Catalog No. 3245MP010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3245MP010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3245MP010 Supplier R&D Systems™ Supplier No. 3245MP010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (Catalog # ES010 ). The specific activity is >75 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O88766
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl and CaCl2.
Gene ID (Entrez) 4317
Molecular Weight (g/mol) M.W. (Observed): 64 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Name MMP-8
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived rat MMP-8 protein Leu21-Pro466, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Pro form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Mmp8
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein MMP-8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.