Learn More
R&D Systems™ Rat PA28 Activator alpha Subunit Recombinant Protein
Description
Specifications
Specifications
| Formulation | Supplied as a solution in HEPES, NaCl, DTT and EDTA. |
| Gene ID (Entrez) | 29630 |
| Molecular Weight (g/mol) | 29 kDa |
| Name | PA28 Activator alpha Subunit |
| Quantity | 100 μg |
| Source | E. coli-derived rat PA28 Activator alpha Subunit protein Ala2-Tyr249 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.