Learn More
R&D Systems™ Recombinant Human MMP-10 Western Blot Standard Protein
Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Gene ID (Entrez) | 4319 |
| Name | MMP-10 Western Blot Standard |
| Quantity | 1 Vial |
| Storage Requirements | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Gene Symbol | MMP10 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Protein | MMP-10 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.