Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MMP-8 Western Blot Standard Protein

Catalog No. WBC017
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WBC017 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. WBC017 Supplier R&D Systems™ Supplier No. WBC017
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot

The Recombinant Human MMP-8 Western Blot Standard Protein has been validated for the following applications: Western Blot.

Specifications

For Use With (Application) Western Blot
Gene ID (Entrez) 4317
Name MMP-8 Western Blot Standard
Quantity 1 Vial
Storage Requirements Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Symbol MMP8
Conjugate Unconjugated
Recombinant Recombinant
Protein MMP-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.