Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X)
SDP

Catalog No. AR011 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AR011 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.
Catalog No. AR011 Supplier R&D Systems™ Supplier No. AR011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.

Optimized for the extended culture of pluripotent stem cell-derived cardiomyocytes. R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X) is nutrient-rich, fully defined, and serum-free.

  • Maintains healthy cardiomyocyte population for longer than 200 days
  • High lot-to-lot consistency decreases experimental variation

Repeated mechanical stress can negatively affect cell attachment and survival. For extended culture, daily media exchanges should be handled as gently as possible.

Specifications

Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Concentration 50 X
Form Liquid
Product Type Maintenance Media Supplement
For Use With (Application) Detection of Cardiac Troponin T in Human ES-derived Cardiomyocytes, Troponin I Expression in Human ES-derived Cardiomyocytes
Product Line R&D Systems
Quantity 10 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.