Description
- Supports reproducible MSC growth in vitro with a formula to reduce variation
- Ready-to-use formula can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
- Has been developed and optimized for use with human, mouse, and rat MSCs
Specifications
Specifications
| Product Type | Stem Cell Expansion Media |
| Product Line | R&D Systems |
| Quantity | 250 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |