Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Osteogenic/Adipogenic Base Media
GSA_VASDP

Catalog No. CCM007 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM007 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM007 Supplier R&D Systems™ Supplier No. CCM007
Add to Cart
Add to Cart
This item is not returnable. View return policy

Base media for the differentiation of MSCs into osteocytes or adipocytes

Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes or adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.

  • Supports reproducible human, mouse, and rat MSC differentiation
  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce osteogenesis or adipogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Osteogenic Supplement (Mfr. Nos. CCM008 or CCM009) or StemXVivo Adipogenic Supplement (Mfr. Nos. CCM011) to reduce variation in differentiation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Stem Cell Media
Quantity 250 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.