Description
Mesenchymal stem/stromal cells (MSCs) are a rare population of multipotent cells that can be derived from a number of tissues such as bone marrow, adipose, and placenta.
- Lacks serum to reduce experimental variation
- Is optimized to support reproducible human MSC growth in vitro
- Can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
- Is suitable for transitioning cultures to pre-clinical expansion protocols
Specifications
Specifications
| Product Type | Stem Cell Expansion Media |
| Product Line | R&D Systems |
| Quantity | 500 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |