Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Xeno-Free Human MSC Expansion Media
SDP

Catalog No. CCM021 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM021 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM021 Supplier R&D Systems™ Supplier No. CCM021

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimized for the expansion of human mesenchymal stem cells

Mesenchymal stem cells are often expanded in media that contain serum and other components that are of animal origin. The presence of animal-derived factors in media can introduce experimental variability and limit the use of the cultured cells in translational or clinical studies.

  • Maintains MSCs in an undifferentiated state
  • Free of animal components that can introduce variability and pose a safety risk
  • Suitable for translational studies

Specifications

Product Type Stem Cell Expansion Media
Product Line R&D Systems
Quantity 500 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.