Learn More
Description
- Results from the chemical conjugation of streptavidin and calf intestinal alkaline phosphatase
- Recommended working dilution is 1:1000 in 1% BSA, 0.05% Tween™ 20, 0.025M Tris, 0.5M NaCl (pH 7.4)
This product contains sodium azide and thimerosal. Take appropriate protective precautions when working with these compounds.
Applications:
ELISA Detection
Specifications
Specifications
| Formulation | Supplied as a solution in 0.1M Tris, 25mM MOPS, 2% polyethylene glycol, 50mM NaCl, 0.5mM Mg2Cl2, 0.005% thimerosal, 0.05% Tween 20, 0.75mM NaN3, 0.015 mM BSA. |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
