Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Streptavidin-Phycoerythrin

Catalog No. F0040
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0040 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F0040 Supplier R&D Systems™ Supplier No. F0040
Add to Cart
Add to Cart
This item is not returnable. View return policy

Applications: Flow Cytometry

  • Secondary reagent in immunofluorescent assays using biotinylated primary labeling reagents
  • 10μg SA-PE in 1mL saline containing up to 0.5% BSA and 0.1% sodium azide
  • R-Phycoerythrin has an absorption spectrum from 480 to 566nm and has optimal emission at 575nm

Applications:

Flow Cytometry

Specifications

Formulation Streptavidin-PE, Ready-to-use Liquid
Quantity 100 Tests
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.