Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TACS TdT In Situ Apoptosis Detection Kit, Blue Label
SDP

Catalog No. 481130K
Change view
Click to view available options
Detection Method:
Blue Label
DAB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
481130K Blue Label
481030K DAB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 481130K Supplier R&D Systems™ Supplier No. 481130K

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Proteinase K Solution; Cytonin™; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Blue Label; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red

A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy

Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using streptavidin-horseradish peroxidase and the colorimetric substrate TACS Blue Label

Specifications

Quantity 30 Tests
Product Type TACS TdT In Situ Apoptosis Detection Kit
Content And Storage Proteinase K, Cytonin, TdT enzyme, dNTPs, StrepHRP, TACS-Blue Label, Blue Streptavidn-Diluent, Cations, TACS-Nuclease, Nuclear Fast Red. The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Line R&D Systems
Detection Method Blue Label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.