Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
SDP
Catalog No. 482830K
Change view
Click to view available options
Detection Method:
Basic
Blue Label
DAB
Replenisher
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
482830K Basic
482830BK Blue Label
482830DK DAB
482830R Replenisher
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 482830K Supplier R&D Systems™ Supplier No. 482830K
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
Specifications
Quantity 30 Tests
Product Type TACS-XL In Situ Apoptosis Detection Kit
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Line R&D Systems
Detection Method Basic