Learn More
R&D Systems™ TACS XTT Cell Proliferation/Viability Assay
Description
Includes
XTT Reagent, 5 x 25mL; XTT Activator, 5 x 0.5mL
XTT is added directly to the culture medium and cleavage of the tetrazolium salt to formazan occurs in metabolically active cells
The orange formazan dye generated can be measured by absorbance at 490nm (or 450nm) in a microplate reader to yield rapid re
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 2500 Tests |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |